missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADAMTSL4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30019568
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019568 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30019568 Supplier Invitrogen™ Supplier No. PA5148417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADAMTSL4 Polyclonal Antibody for ICC/IF

This gene is a member of ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs)-like gene family and encodes a protein with protein with seven thrombospondin type 1 repeats. The thrombospondin type 1 repeat domain is found in many proteins with diverse biological functions including cellular adhesion, angiogenesis, and patterning of the developing nervous system. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTSL4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene ADAMTSL4
Gene Accession No. Q6UY14
Gene Alias a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 4; adamtsl4; ADAMTSL-4; adamtsl4 {ECO:0000312; ADAMTS-like 4; ADAMTS-like protein 4; ECTOL2; EMBL:AAH99119.1}; metalloendopeptidases; PP1396; thrombospondin repeat containing 1; thrombospondin repeat protein 1; thrombospondin repeat-containing protein 1; Tsrc1; UNQ2803/PRO34012
Gene Symbols ADAMTSL4
Host Species Rabbit
Immunogen E. coli-derived Human ADAMTSL4 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54507
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.