missing translation for 'onlineSavingsMsg'
Learn More

ADAM5 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7209418 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16841685 20 μL 20µL
16831685 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16841685 Supplier Proteintech Supplier No. 221471AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is specific to ADAM5.

ADAM5 (putative disintegrin and metalloproteinase domain-containing protein 5) is also named as ADAM5P, TMDC2(tMDCII). The tMDC II, abundantly expressed, sperm surface MDC proteins identified in rodent and macaque systems, is also nonfunctional in the human. It has 7 isoforms produced by alternative splicing. It is expressed in testis highly by northern blot.

Specifications

Antigen ADAM5
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ADAM5
Gene Accession No. Q6NVV9
Gene Alias ADAM5, ADAM5P, TMDCII
Gene Symbols ADAM5
Host Species Rabbit
Immunogen ADAM5 Fusion Protein Ag17590
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255926
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.