missing translation for 'onlineSavingsMsg'
Learn More

ADAM28 Antibody (1004266) [DyLight 350], Novus Biologicals™

Product Code. 18553659 Shop All Bio Techne Products
Cambia vista
Click to view available options
Quantity:
0.1 mL
Dimensione della confezione:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
18553659 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18553659 Fornitore Novus Biologicals N. del fornitore FAB93252D

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

ADAM28 Monoclonal antibody specifically detects ADAM28 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifica

Antigen ADAM28
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1004266
Conjugate DyLight 350
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human ADAM28, Leu72 & Leu191-Ala623,, Accession # Q0UKQ2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10863
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.