missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACVR2B Polyclonal Antibody
GREENER_CHOICE

Product Code. 13402347
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13402347 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13402347 Supplier Invitrogen™ Supplier No. PA528231

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, SK-N-SH, IMR-32, SK-N-AS, mosue cerebellum, mouse testis, ACVR2B-transfected 293T. Predicted reactivity: Mouse (99%), Rat (99%), Pig (100%), Chicken (89%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ACVR2B is a type II member of the TGF-beta family of receptor ser/thr kinases and is involved in the activin and myostatin signaling pathway. Activins and inhibins are members of the TGF-beta superfamily due to amino acid homology with respect to the conservation of 7 of the 9 cysteine residues common to all TGF-beta forms. To date, seven type I and five type II activin receptors have been cloned from mammals, including activin receptor IA, activin receptor IIA, activin receptor IB, and activin receptor IIB. In addition, two splice variants of activin receptor IIA and five splice variants of activin receptor IIB have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ACVR2B
Gene Accession No. P27040, Q13705
Gene Alias activin A receptor type 2B; activin A receptor type IIB; activin A receptor, type IIB; activin receptor IIB; activin receptor type IIB; Activin receptor type-2B; Activine receptor 2b (transmembrane serine kinase); ActRIIB; ACTR-IIB; ActRIIB type II activin receptor B; ACVR2B; HTX4; MGC116908
Gene Symbols ACVR2B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Activin Receptor Type IIB. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11481, 93
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.