Learn More
ACTR2 Antibody (S04-3H3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19646-100ul
 View more versions of this product
Product Code. 18636973
  • 645.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
ACTR2 Monoclonal antibody specifically detects ACTR2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACTR2
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S04-3H3
Unconjugated
actin-related protein 2, ARP2 (actin-related protein 2, yeast) homolog, ARP2 actin-related protein 2 homolog (yeast), ARP2Actin-like protein 2
A synthetic peptide of human ACTR2 (Uniprot # P61160)
100 μg
Protein Kinase
10097
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.