missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTR1A Monoclonal Antibody (3E5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520935
This item is not returnable.
View return policy
Description
Peptide Sequence: LVFAIQKSDM DLRRTLFSNI VLSGGSTLFK GFGDRLLSEV KKLAPKDVKI RISAPQERLY STWIGGSILA SLDTFKKMWV SKKEYEEDGA RSIHRKT.
This gene encodes a 42.6 kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 8-13 copies per dynactin molecule, and is the most abundant molecule in the dynactin complex. It is an actin-related protein, and is approximately 60% identical at the amino acid level to conventional actin.
Specifications
| ACTR1A | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P61163 | |
| ACTR1A | |
| ACTR1A (NP_005727, 279 a.a. ~ 375 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 3E5 | |
| Unconjugated | |
| ACTR1A | |
| actin related protein 1A; actin-related protein 1 alpha; actin-related protein 1 homolog A; actin-RPV; ACTR1A; actr1a {ECO:0000250; alpha-Arp1; alpha-centractin; ARP1; ARP1 actin related protein 1 homolog A; ARP1 actin-related protein 1 homolog A, centractin alpha; ARP1 actin-related protein 1 homolog A, centractin alpha (yeast); ARP1 actin-related protein 1A, centractin alpha; Arp1 centractin alpha; Arp1 contractin alpha; Arp1A; centractin; Centrosome-associated actin homolog; CTRN1; FLJ52695; FLJ52800; FLJ55002; UniProtKB:P61163}; yeast homolog A | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10121 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction