missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Mouse, Clone: r57, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 5443-MSM6-P1
Additional Details : Weight : 0.01000kg
Specifications
ACTH | |
Cytoplasm and Secreted | |
r57 | |
Unconjugated | |
NP_001306133 | |
Mouse | |
ACTH is ∼5kDa, and the POMC precursor is ∼30kDa. The molecular weight of POMC depends upon isoform variation and post-translational modifications. | |
Protein A/G Purified | |
RUO | |
5443 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
POMC | |
N-terminal fragment of human ACTH conjugated to KLH | |
Normal pituitary gland or pituitary tumor. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |