Learn More
Acrylic Acid-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Acrylic Acid-D4 |
| CAS | 285138-82-1 |
| Molecular Formula | C3 D4 O2 |
| Quantity | 2.5 mg |
| Synonym | 2-Propenoic-2,3,3-d3 acid-d (9CI), Deuterio 2,3,3-trideuterioprop-2-enoate, Acrylic Acid-d4, Acrylic Acid-D4, 2-Propenoic acid-d4 |
| InChI Formula | InChI=1S/C3H4O2/c1-2-3(4)5/h2H,1H2,(H,4,5)/i1D2,2D/hD |
| SMILES | [2H]OC(=O)C(=C([2H])[2H])[2H] |
| IUPAC Name | deuterio 2,3,3-trideuterioprop-2-enoate |
| Molecular Weight (g/mol) | 76.0873 |
| Formula Weight | 76.0462 |