Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Posidyne Membrane Optimized for Scale Up
Product Code. 17124451
Change view
Click to view available options
:
0.2 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17124451 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17124451 Supplier Cytiva (formerly Pall Lab) Supplier No. 4908
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Syringe filters ideal for biological and general research.
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Posidyne
Pore Size 0.2 μm
Sterility Sterile