Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PVDF Membrane
Syringe Filter with hydrophilic membrane compatible with a wide variety of solvents.
Brand:  Cytiva (formerly Pall Lab) AP-4792
Additional Details : Weight : 1.13400kg
 View more versions of this product
Product Code. 17174691
  • 870.00 € / Pack of 200
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
  • Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
  • For research use only
Specifications
Specifications
Glass Fiber, PVDF
Non-sterile
<125 μL
200/Cs.
82°C
0.2 μm
3.9 cm2
Polypropylene
25/AutoPack Tubes
25 mm