Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PVDF Membrane
Syringe Filter with hydrophilic membrane compatible with a wide variety of solvents.
Brand: Cytiva (formerly Pall Lab) AP-4309
Additional Details : Weight : 0.10000kg
Beschreibung
- Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
- For research use only
Spezifikation
Glass Fiber, PVDF | |
Non-sterile | |
<125 μL | |
200/Cs. | |
82°C |
0.45 μm | |
3.9 cm2 | |
Polypropylene | |
25/AutoPack Tubes | |
25 mm |