Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PVDF Membrane
Syringe Filter with hydrophilic membrane compatible with a wide variety of solvents.
Brand:  Cytiva (formerly Pall Lab) AP-4797
Additional Details : Weight : 3.01150kg
 View more versions of this product
Product Code. 17114701
  • 3250.00 € / Pack of 1000
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Description
Description
  • Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
  • For research use only
Specifications
Specifications
PVDF
Non-sterile
<100 μL
1000/Pk.
25 mm
0.2 μm
3.9 cm2
Polypropylene
82°C