Learn More
Acid Blue 45 (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Acid Blue 45 (Technical Grade) |
| CAS | 2861-02-1 |
| Molecular Formula | C14H8N2O10S2Na2 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C14H10N2O10S2.2Na/c15-3-1-5(27(21,22)23)11(17)9-7(3)13(19)10-8(14(9)20)4(16)2-6(12(10)18)28(24,25)26;;/h1-2,17-18H,15-16H2,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2 |
| SMILES | [Na+].[Na+].Nc1cc(c(O)c2C(=O)c3c(N)cc(c(O)c3C(=O)c12)S(=O)(=O)[O-])S(=O)(=O)[O-] |
| IUPAC Name | disodium;4,8-diamino-1,5-dihydroxy-9,10-dioxoanthracene-2,6-disulfonate |
| Molecular Weight (g/mol) | 474.33 |
| Formula Weight | 473.9416 |