missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-L-glutamine, TRC

Product Code. 30322800
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 30322800

Brand: Toronto Research Chemicals TRCA297860500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-L-glutamine
CAS 2490-97-3
Molecular Formula C7H12N2O4
Synonym L-Glutamine, N2-acetyl- (9CI, ACI), Glutamine, N2-acetyl-, L- (6CI, 7CI, 8CI), N2-Acetyl-L-glutamine (ACI), (2S)-2-Acetamido-5-amino-5-oxopentanoic acid, (2S)-4-Carbamoyl-2-acetamidobutanoic acid, Aceglutamid, Aceglutamide, Acetylglutamine, Acutil S, N2-Acetylglutamine, NSC 186896, α-N-Acetyl-L-glutamine
InChI Formula InChI=1S/C7H12N2O4/c1-4(10)9-5(7(12)13)2-3-6(8)11/h5H,2-3H2,1H3,(H2,8,11)(H,9,10)(H,12,13)/t5-/m0/s1
SMILES CC(=O)N[C@@H](CCC(=O)N)C(=O)O
IUPAC Name (2S)-2-acetamido-5-amino-5-oxo-pentanoic acid
Molecular Weight (g/mol) 188.18
Quantity 500 mg
Formula Weight 188.0797
Shipping Condition Room Temperature
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.