Learn More
Acetyl Coenzyme A Lithium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Acetyl Coenzyme A Lithium Salt |
| CAS | 32140-51-5 |
| Molecular Formula | C23H38N7O17P3S . Lix |
| Quantity | 5 mg |
| Synonym | Lithium acetyl-CoA, Coenzyme A, S-acetate, lithium salt |
| InChI Formula | InChi=1S/C23H38N7O17P3S.3Li/c1-12(31)51-7-6-25-14(32)4-5-26-21(35)18(34)23(2,3)9-44-50(41,42)47-49(39,40)43-8-13-17(46-48(36,37)38)16(33)22(45-13)30-11-29-15-19(24)27-10-28-20(15)30;;;/h10-11,13,16-18,22,33-34H,4-9H2,1-3H3,(H,25,32)(H,26,35)(H,39,40)(H,41,42)(H2,24,27,28)(H2,36,37,38);;;/q;3*+1/p-3/t13-,16-,17-,18+,22-;;;/m1.../s1 |
| SMILES | NC1=C2C(N([C@H]3[C@H](O)[C@H](OP(O)(O)=O)[C@@H](COP(OP(O)(OCC([C@@H](O)C(NCCC(NCCSC(C)=O)=O)=O)(C)C)=O)(O)=O)O3)C=N2)=NC=N1.[Li] |
| IUPAC Name | Lithium S-(2-(3-((2R)-4-(((((((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-hydroxy-3-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)(hydroxy)phosphoryl)oxy)-2-hydroxy-3,3-dimethylbutanamido)propanamido)ethyl) ethanethioate |
| Molecular Weight (g/mol) | 816.5 |