missing translation for 'onlineSavingsMsg'
Learn More

Acetobromo-alpha-D-glucose, 97%, stab. with 2% calcium carbonate

Product Code. p-7044030
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11316816

Brand: Thermo Scientific Alfa Aesar L04151.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 572-09-8 | C14H19BrO9 | 411.20 g/mol

Acetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.

Solubility
Soluble in ether, chloroform, ethanol, acetone, dichloromethane, ether, ethyl, methanol, and tetrahydrofuran. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 572-09-8
Molecular Formula C14H19BrO9
Molecular Weight (g/mol) 411.20
MDL Number MFCD00063254
InChI Key CYAYKKUWALRRPA-HAGGZJDPNA-N
Synonym 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, acetobromglucose, acetobromoglucose, acetobromo-alpha-d-glucose, a-acetobromglucose, acetobromo glucose, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, stab. with calcium carbonate
PubChem CID 101776
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

Specifications

Melting Point 84°C to 89°C
Quantity 10 g
Beilstein 96669
Sensitivity Moisture sensitive
Merck Index 14,60
Solubility Information Soluble in ether,chloroform,ethanol,acetone,dichloromethane,ether,ethyl,methanol,and tetrahydrofuran. Insoluble in water.
Optical Rotation +198° (c=3 in Chloroform)
Formula Weight 411.22
Percent Purity 97%
Chemical Name or Material Acetobromo-alpha-D-glucose, stabilized with 2% calcium carbonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.