Learn More
Acetaminophen-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Acetaminophen methyl-d3 |
| CAS | 60902-28-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 2H3 H6 N O2 |
| Quantity | 10 mg |
| Synonym | Acetaminophen D3 (Methyl D3), Acetamide-2,2,2-d3, N-(4-hydroxyphenyl)-, N-(4-Hydroxyphenyl)acetamide-2,2,2-d3, Paracetamol D3, Paracetamol D3 (Methyl D3) |
| InChI Formula | InChI=1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)/i1D3 |
| SMILES | [2H]C([2H])([2H])C(=O)Nc1ccc(O)cc1 |
| IUPAC Name | 2,2,2-trideuterio-N-(4-hydroxyphenyl)acetamide |
| Molecular Weight (g/mol) | 154.18 |
| Show More |