Learn More
Cole-Parmer™ Acetal Valved Quick-Disconnect Hose Barb Insert
12.95 € - 15.75 €
Product Code | Brand | Overall Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Overall Length (Metric) | Price | Quantity & Availability | |||||
11740289
|
Cole-Parmer™
06360-83 |
7.8 cm |
12.95 €
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
11735149
|
Cole-Parmer™
06360-43 |
4.8 cm |
15.75 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Specifications
Autoclavable | |
Acetal | |
1 Pack |