missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACER2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101415
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ASAH3L.
The sphingolipid metabolite sphingosine-1-phosphate promotes cell proliferation and survival, whereas its precursor, sphingosine, has the opposite effect. The ceramidase ACER2 hydrolyzes very long chain ceramides to generate sphingosine.
Specifications
| ACER2 | |
| Polyclonal | |
| Unconjugated | |
| ACER2 | |
| 2410116I05Rik; ACER2; Acylsphingosine deacylase 3-like; alkaline CDase 2; alkaline ceramidase 2; AlkCDase 2; ALKCDase2; Asah3l; cancer related gene-liver 1 (CRG-L1); cancer-related gene liver 1 protein; ceramide hydrolase; CRG-L1; haCER2; maCER2; N-acylsphingosine amidohydrolase 3-like; PP11646 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 230379, 340485 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5QJU3, Q8VD53 | |
| ACER2 | |
| A synthesized peptide derived from human ACER2(Accession Q5QJU3), corresponding to amino acid residues Y209-P259. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction