Learn More
Aceclofenac-d4 (phenyl-d4-acetoxy), CDN
Specifications
Specifications
| Chemical Name or Material | Aceclofenac-D4 (2-aminophenyl-D4) |
| CAS | 2748492-20-6 |
| Molecular Formula | C16 D4 H9 Cl2 N O4 |
| Quantity | 0.005 g |
| Synonym | Aceclofenac-d4 (phenyl-d4-acetoxy), Aceclofenac-d4, Aceclofenac-D4 (2-aminophenyl-D4), 2-[(2,6-Dichlorophenyl)amino]benzene-3,4,5,6-d4-acetic acid carboxymethyl ester, 2-[[2-[(2,6-Dichlorophenyl)amino]phenyl-3,4,5,6-d4]acetoxy]acetic acid |
| InChI Formula | InChI=1 S/C16H13Cl2NO4/c17-11-5-3-6-12(18)16(11)19-13-7-2-1-4-10(13)8-15(22)23-9-14(20)21/h1-7,19 H,8-9H2,(H,20,21)/i1D,2 D,4 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Nc2c(Cl)cccc2Cl)c(CC(=O)OCC(=O)O)c1[2 H] |
| IUPAC Name | 2-[2-[2,3,4,5-tetradeuterio-6-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid |
| Molecular Weight (g/mol) | 358.2094 |
| Formula Weight | 357.0473 g/mol |
| Show More |