Learn More
Miele Professional™ Closed Plinth UG 30-90/60-85
Product Code. 15457724 Shop All Miele Products
Click to view available options
:
plinth; Dimensions (D x W): 60 x 90mm
Product Code. 15457724
Brand: Miele Professional™ 12650160_OLD
Only null left
Add to basket
Add to basket
Closed plinth UG 30-90/60-85 to raise the machine incl. side unit, 60 cm deep, 90 cm wide
For use in Aqua Purificator PG 8595 and DOS cabinet PG 8596
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Product Type Plinth/Base
Color Stainless Steel
For Use With (Equipment) Large-capacity washer-disinfectors, laboratory