Learn More
Cytiva Accessories for SE 250/260 Mini-vertical Gel Units:Glass Plate
Brand: Cytiva SE202GN-5
214.70 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.15900kg
Specifications
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit | |
Glass | |
10 cm |
Glass Plate | |
Rectangular | |
8 cm |