missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAA2 Antibody (OTI1C10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71602CL1
This item is not returnable.
View return policy
Description
ACAA2 Monoclonal antibody specifically detects ACAA2 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| ACAA2 | |
| Monoclonal | |
| CoraFluor 1 | |
| 3-ketoacyl-CoA thiolase, mitochondrial, Acetyl-CoA acyltransferase, acetyl-CoA acyltransferase 2, acetyl-Coenzyme A acyltransferase 2, beta ketothiolase, beta-ketothiolase, DSAEC, EC 2.3.1, EC 2.3.1.16, FLJ35992, FLJ95265, Mitochondrial 3-oxoacyl-CoA thiolase, mitochondrial 3-oxoacyl-Coenzyme A thiolase, T1 | |
| Full length human recombinant protein of human ACAA2(NP_006102) produced in HEK293T cell. | |
| 0.1 mL | |
| Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 10449 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI1C10 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction