Learn More
AC710, MedChemExpress
Product Code. 16496196
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16496196
Brand: Medchem Express HY13493
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress AC710 is a potent PDGFR inhibitor with Kds of 0.6, 1.57, 1, 1.3, 1.0 nM for FLT3, CSF1R, KIT, PDGFRα and PDGFRβ, respectively.
AC710 is a potent PDGFR inhibitor with Kds of 0.6, 1.57, 1, 1.3, 1.0 nM for FLT3, CSF1R, KIT, PDGFRα and PDGFRβ, respectively.
Specifications
Chemical Name or Material AC710
CAS 1351522-04-7
Color White
Purity Grade Notes Research
Molecular Formula C31H42N6O4
Quantity 100 mg
Solubility Information DMSO : 14 mg/mL (24.88 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC=C(OC2CC(C)(C)N(CC)C(C)(C)C2)C=C1)NC3=CC=C(NC(NC4=NOC(C(C)(C)C)=C4)=O)C=C3
Molecular Weight (g/mol) 562.7
Formula Weight 562.7
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease