Learn More
Ac-Gly-BoroPro, MedChemExpress
Product Code. 16420909
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 16420909
Brand: Medchem Express HY101801
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Ac-Gly-BoroPro is a selective FAP inhibitor with a Ki of 23 nM.
Ac-Gly-BoroPro is a selective FAP inhibitor with a Ki of 23 nM.
Specifications
Chemical Name or Material Ac-Gly-BoroPro
Physical Form Solid
CAS 886992-99-0
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C8H15BN2O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (233.61 mM)
SMILES O=C(N1[C@@H](B(O)O)CCC1)CNC(C)=O
Molecular Weight (g/mol) 214.03
Formula Weight 214.03
Percent Purity 98.0%
Grade Research