missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ac-dC Phosphoramidite, standard grade, serum vial bottle

Product Code. 18024110 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18024110 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18024110 Supplier Thermo Scientific™ Supplier No. 29172707

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ac-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis.

Ac-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. By using Ac-dC, Bz-dA, and DMF-dG monomers, fast cleavage and deprotection can be effected by a 1:1 mixture of aqueous ammonium hydroxide and aqueous methylamine (or AMA) in 10 min. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:

  • Full chemical name – 5'-O-(4,4'-Dimethoxytrityl)-N4-Acetyl-2'-deoxycytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)-phosphoramidite
  • Molecular formula – C41H50N5O8P
  • Base – dC
  • Base Protection – Ac
  • Molecular weight – 771.8 g/mol
  • CAS number – 154110-40-4
  • HPLC Purity – ≥98%
  • 31P NMR Purity – ≥98%
TRUSTED_SUSTAINABILITY

Specifications

Format Powder
For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type Ac-dC CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxycytidine (dC)
Reagent Features Fast deprotection
Shipping Condition Room Temperature
Purity 98%
Quantity 2 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.