Learn More
Brady™ AC Adaptor
Click to view available options
Description:
AC Adaptor, Europe 220 V
AC Adaptor, UK 240 V
Unit Size:
Each
Description
- 800747: AC Adapter Europe (for ABC or KEY printers).
- 710612: Accessory for the BMP™41/51/53/61/71 Label Printers.
- 710613: Accessory for the BMP™51/53 Label Makers.
Specifications
Specifications
| Product Type | AC Adapter |
| For Use With (Equipment) | BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer, BMP71 Label Printer, BradyPrinter M611 |
| Description | AC Adaptor, UK 240 V |
| Quantity | 1 Ea. |
| Voltage | 240 V |
| Alternate Part Number | M41/50/61/71/611-AC-BC-UK |