Learn More
Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes
Shop All Thermo Scientific ProductsDescription
Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes.
Maximum specificity—Chemically-modified Thermo-Start™ Taq remains completely inactive until heated to 95°C for 15 minutes, thereby preventing nonspecific amplification
Improved efficiency—The use of dTTP in place of dUTP increases reaction efficiency by approximately 5% and therefore reduces reaction time
Increased sensitivity—The combination of Thermo-Start Taq and dTTP increases sensitivity and allows for detection of low copy number templates
Instrument specific—Individually optimized qPCR mixes designed for all major qPCR / Real-Time PCR instruments
Reproducible results—All mixes undergo extensive quality control testing to ensure batch-to-batch consistency
Simple protocol—Master mix format reduces protocol steps and minimizes risk of pipetting errors and sample contamination
Includes:
ABsolute qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, ROX™ passive reference dye (only in ROX mixes); Separate ROX vial (only in kits indicated)
Recommended for:
Ideal for qPCR analysis in molecular biology applications, including gene expression and SNP typing
Specifications
Specifications
| Content And Storage | ABsolute qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, ROX™ passive reference dye (only in ROX mixes); Separate ROX vial (only in kits indicated). Store at -20°C. Avoid freeze/thaw cycles |
| Description | ABsolute QPCR Capillary Mix, 200 rxns |
| Quantity | 250 x 20µL rxn |
| For Use With (Application) | RT-PCR |
| Activation Time | heated to 95°C for 15 minutes |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.