Learn More
Abridin, TRC
Specifications
Specifications
| Chemical Name or Material | Abridin |
| CAS | 73030-56-5 |
| Molecular Formula | C25H36O2 |
| Quantity | 25 mg |
| Synonym | (22E)-26,27-Dinorcholesta-4,22-diene-3,24-dione |
| InChI Formula | InChI=1S/C25H36O2/c1-16(5-6-17(2)26)21-9-10-22-20-8-7-18-15-19(27)11-13-24(18,3)23(20)12-14-25(21,22)4/h5-6,15-16,20-23H,7-14H2,1-4H3/b6-5+/t16-,20+,21-,22+,23+,24+,25-/m1/s1 |
| SMILES | C[C@@H](\C=C\C(=O)C)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(E,2R)-5-oxohex-3-en-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 368.55 |