Learn More
Novus Biologicals™ AbPure™ Antibody Concentration and Clean-Up Antibody Purification Kit
Shop All Bio Techne ProductsDescription
Kit Components
- 3 spin cartridge/collecting tube assemblies
- 1 bottle of AbPure Conjugation Buffer
Specifications
Specifications
| Content And Storage | 4°C |
| Quantity | 3 Tests |
| Kit Contents | 3 spin cartridge/collecting tube assemblies, 1 bottle of AbPure Conjugation Buffer |
| Sufficient For | 3 columns |
| Product Type | AbPure Antibody Concentration and Clean-Up Antibody Purification Kit |