missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABL1 [Q252H] Recombinant Human Protein, Invitrogen™
Human ABL1, internal fragment, amino acids P118-S535 (as in GenBank entry NM_005157.3), Q252H mutant, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
492.00 € - 7490.00 €
Specifications
For Use With (Application) | Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation |
---|---|
Formulation | 50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol |
Molecular Weight (g/mol) | 76.2kDa |
Storage Requirements | Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
Gene Alias | c-Abl, JTK7, p150 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458823
|
Invitrogen
PV6155 |
100μg |
1678.00 €
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15448823
|
Invitrogen
PV6154 |
10μg |
492.00 €
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468823
|
Invitrogen
PV6156 |
1mg |
7490.00 €
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
76.2kDa | |
c-Abl, JTK7, p150 | |
Human | |
GST Tag, His Tag |
50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 118-535 | |
Recombinant |