missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-132PE
This item is not returnable.
View return policy
Description
ABCG1 Polyclonal antibody specifically detects ABCG1 in Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
| ABCG1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit | |
| Purified |
| Flow Cytometry | |
| PE | |
| ABC transporter 8, ABC8, ABCG1, ATP-binding cassette sub-family G member 1, ATP-binding cassette transporter 8, ATP-binding cassette transporter member 1 of subfamily G, ATP-binding cassette, sub-family G (WHITE), member 1, homolog of Drosophila white, MGC34313, White protein homolog, white protein homolog (ATP-binding cassette transporter 8), WHITE1, WHT1 | |
| A synthetic peptide made to an internal region of human ABCG1 (between residues 300-400). [UniProt# P45844] | |
| 0.1 mL | |
| ABC Transporters, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 9619 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction