missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104371
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ABCD1.
This gene is a member of the UBA domain family, whose members include proteins having connections to ubiquitin and the ubiquitination pathway. The ubiquitin associated domain is thought to be a non-covalent ubiquitin binding domain consisting of a compact three helix bundle. This particular protein originates from a gene locus in a refined region on chromosome 9 undergoing loss of heterozygosity in nasopharyngeal carcinoma (NPC). Taking into account its cytogenetic location, this UBA domain family member is being studies as a putative target for mutation in nasopharyngeal carcinomas. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ABCD1 | |
| Polyclonal | |
| Unconjugated | |
| ABCD1 | |
| ABC42; ABCD1; Adrenoleukodystrophy protein; ALD; Aldgh; ALDP; AMN; ATP binding cassette subfamily D member 1; ATP-binding cassette sub-family D member 1; ATP-binding cassette, subfamily D (ALD), member 1; ATP-binding cassette, sub-family D (ALD), member 1; ATP-binding cassette, sub-family D, member 1; RGD1562128; X-linked adrenoleukodystrophy (ALD) gene homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 215 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P33897 | |
| ABCD1 | |
| A synthesized peptide derived from human ABCD1(Accession P33897), corresponding to amino acid residues Q544-G594. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction