missing translation for 'onlineSavingsMsg'
Learn More

ABCC11 Antibody (ABCC11/2438), Novus Biologicals™

Product Code. 18662464 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18662464 20 μg 20µg
18613685 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18662464 Supplier Novus Biologicals Supplier No. NBP32390220ug
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABCC11 Monoclonal antibody specifically detects ABCC11 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCC11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ABCC11/2438
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias ATP-binding cassette protein C11, ATP-binding cassette transporter MRP8, ATP-binding cassette transporter sub-family C member 11, ATP-binding cassette, sub-family C (CFTR/MRP), member 11, EWWD, MRP8ATP-binding cassette sub-family C member 11, Multidrug resistance-associated protein 8, multi-resistance protein 8, WW
Host Species Mouse
Immunogen Recombinant fragment (around aa400-600) of human ABCC11 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85320
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.