missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCC11 Antibody (ABCC11/2438), Novus Biologicals™
Mouse Monoclonal Antibody
245.00 € - 504.00 €
Specifications
| Antigen | ABCC11 |
|---|---|
| Clone | ABCC11/2438 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613685
|
Novus Biologicals
NBP3-23902-100ug |
100 μg |
504.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662464
|
Novus Biologicals
NBP3-23902-20ug |
20 μg |
245.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABCC11 Monoclonal antibody specifically detects ABCC11 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ABCC11 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| ATP-binding cassette protein C11, ATP-binding cassette transporter MRP8, ATP-binding cassette transporter sub-family C member 11, ATP-binding cassette, sub-family C (CFTR/MRP), member 11, EWWD, MRP8ATP-binding cassette sub-family C member 11, Multidrug resistance-associated protein 8, multi-resistance protein 8, WW | |
| Recombinant fragment (around aa400-600) of human ABCC11 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| ABCC11/2438 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 85320 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title