Learn More
Abcam VHH Single Domain Anti-Rabbit IgG Fc (HRP), 250 uL
Product Code. 32116377 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μL
Unit Size:
250µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116377 250 μL 250µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32116377 Supplier Abcam Supplier No. ab191866250uL
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Monoclonal antibody, suitable for IHC-P, WB, ELISA, with over 100 citations.
  • Clonality: Monoclonal
  • Applications: IHC-P, WB, ELISA
  • Species Reactivity: Rabbit
  • Storage Buffer: pH: 7.4, Preservative: 0.1% Proclin 300 Solution, Constituents: Sodium phosphate, Sodium chloride, PBS, 30% Glycerol (glycerin, glycerine), 1% BSA
  • Form: Liquid
  • Purification: Affinity purification His Tag
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GD001
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.1% Proclin 300 Solution, Constituents: Sodium phosphate, Sodium chloride, PBS, 30% Glycerol (glycerin, glycerine), 1% BSA
Gene Rabbit IgG
Purification Method Affinity purification His Tag
Quantity 250 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Rabbit.
Target Species Rabbit
Product Type Monoclonal Antibody
Form Liquid
Shipping Condition Blue Ice
Reactivity abrv Rb
Target Molecule Rabbit IgG, Rabbit
Validated Application abrv IHC-P, WB, ELISA
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.