Description
- Host Species: Rabbit
- Clonality: Monoclonal
- Isotype: IgG
- Applications: IP, Flow Cyt, WB, ICC/IF
- Species Reactivity: Mouse, Rat and Human
- Storage Buffer: pH: 7.2 - 7.4, Constituents: PBS
- Form: Liquid
- Purification: Affinity purification Protein A
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Antigen | ATM |
| Applications | Immunoprecipitation, Flow Cytometry, Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | EPR17059 |
| Conjugate | Unconjugated |
| Formulation | pH: 7.2 - 7.4, Constituents: PBS |
| Gene | ATM |
| Gene Symbols | ATM |
| Host Species | Rabbit |
| Purification Method | Affinity purification Protein A |
| Show More |