Learn More
Motic™ Abbe Condenser N.A. 0.90/1.25
Product Code. 15455202
Change view
Click to view available options
For Use With (Equipment):
BA310 and BA410 Series Upright Compound Microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15455202 BA310 and BA410 Series Upright Compound Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15455202 Supplier Motic™ Supplier No. 1101001901581
  • ONLINE EXCLUSIVE PRICE
    67.80 €
    60.51 € / Pack of 1
    Save 7.29 €
    10% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Constructed with a high quality plastic surround on metal inner, optic-holding parts. Motic™ Abbe Condenser, N.A. 0.90/1.25 has a 6-blade diaphragm.
  • Abbe-type condenser.
  • High-quality construction (plastic/metal).
  • Includes a 6-blade diaphragm.
  • Includes a slot for sliders.
Specifications
Type Condenser
Description N.A. 0.90/1.25
For Use With (Equipment) BA310 and BA410 Series Upright Compound Microscopes
Includes Six-blade diaphragm, slot for sliders