missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abarelix Acetate, MedChemExpress
Description
Abarelix Acetate (PPI 149 Acetate
Specifications
Specifications
| Chemical Name or Material | Abarelix Acetate |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C74H99ClN14O16 |
| Quantity | 10 mg |
| Synonym | PPI 149 AcetateR 3827 Acetate |
| Solubility Information | DMSO : ≥ 100 mg/mL (67.75 mM) ∣H2O : 25 mg/mL (16.94 mM; Need ultrasonic) |
| SMILES | CC(O)=O.C[C@H](C(N)=O)NC([C@H]1N(C([C@H](CCCCNC(C)C)NC([C@H](CC(C)C)NC([C@@H](CC(N)=O)NC([C@H](CC2=CC=C(C=C2)O)N(C([C@H](CO)NC([C@@H](CC3=CC=CN=C3)NC([C@@H](CC4=CC=C(Cl)C=C4)NC([C@@H](CC5=CC=C6C=CC=CC6=C5)NC(C)=O)=O)=O)=O)=O)C)=O)=O)=O)=O)CCC1)=O |
| Molecular Weight (g/mol) | 1476.12 |
| Formula Weight | 1476.12 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?