missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AAK1, GST Tag Recombinant Protein
This AAK1 Recombinant Human Protein (1-465) was expressed in insect cells.
Brand: Invitrogen™ A32880
Additional Details : Weight : 0.01000kg
Description
- This AAK1 recombinant human protein (1–465) was expressed in insect cells
- AAK1 serine/threonine kinase is a member of the NAK family and plays a regulatory role in endocytosis
- The purified catalytic domain (1-510) of AAK1 can be used in a number of different applications, but has been specifically validated for use with our LanthaScreen™ Kinase binding assay technology to identify novel kinase inhibitors
- AAK1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Specifications
AAH02695 | |
77.3 kDa | |
NAK Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
100 μg | |
aa 1 - 510 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Baculovirus | |
Recombinant, Catalytic |