missing translation for 'onlineSavingsMsg'
Learn More

AA26-9, MedChemExpress

Product Code. 16457518
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16457518

Brand: Medchem Express HY18522

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AA26-9 is a potent and broad spectrum serine hydrolase inhibitor. AA26-9 targets included serine peptidases, lipases, amidases, esterases, and thioesterases. AA26-9 shows inhibitory activity against approximately 1/3 of the 40+ serine hydrolases detected in immortalized T cell lines .

AA26-9 is a potent and broad spectrum serine hydrolase inhibitor. AA26-9 targets included serine peptidases, lipases, amidases, esterases, and thioesterases. AA26-9 shows inhibitory activity against approximately 1/3 of the 40+ serine hydrolases detected in immortalized T cell lines .

Specifications

Chemical Name or Material AA26-9
Physical Form Solid
CAS 1312782-34-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C7H10N4O
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 100 mg/mL (601.76 mM; Need ultrasonic)
SMILES O=C(N1N=NC=C1)N2CCCC2
Molecular Weight (g/mol) 166.18
Formula Weight 166.18
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.