missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A939572, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
Description
A939572 is a potent, and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 values of mSCD1 and hSCD1, respectively.
Specifications
Specifications
| Chemical Name or Material | A939572 |
| CAS | 1032229-33-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H22ClN3O3 |
| Quantity | 2 mg |
| Solubility Information | DMSO : 100 mg/mL (257.82 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C(N1CCC(OC2=C(Cl)C=CC=C2)CC1)NC3=CC=CC(C(NC)=O)=C3 |
| Molecular Weight (g/mol) | 387.86 |
| Formula Weight | 387.86 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Partager une correction de contenu