missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A-RAF Rabbit anti-Human, Clone: 4U9L7, Novus Biologicals™
Rabbit Monoclonal Antibody
190.00 € - 496.00 €
Specifications
| Antigen | A-RAF |
|---|---|
| Clone | 4U9L7 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:10-1:20 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18381507
|
Novus Biologicals
NBP3-16677-20UL |
20 μg |
190.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18315552
|
Novus Biologicals
NBP3-16677-100UL |
100 μg |
496.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A-RAF Monoclonal antibody specifically detects A-RAF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| A-RAF | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:10-1:20 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ARAF1A-RAF, EC 2.7.11, EC 2.7.11.1, Oncogene ARAF1, PKS, PKS2A-Raf proto-oncogene serine/threonine-protein kinase, Proto-oncogene A-Raf, Proto-oncogene A-Raf-1, Proto-oncogene Pks, RAFA1, Ras-binding protein DA-Raf, serine/threonine-protein kinase A-Raf, v-raf murine sarcoma 3611 viral oncogene homolog, v-raf murine sarcoma 3611 viral oncogene homolog 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 166-290 of human A-RAF (P10398). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 4U9L7 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 369 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title