missing translation for 'onlineSavingsMsg'
Learn More

A-RAF Antibody (OTI2G9), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18132245 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18132245 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18132245 Supplier Novus Biologicals Supplier No. NBP270201JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

A-RAF Monoclonal specifically detects A-RAF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A-RAF
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G9
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ARAF1A-RAF, EC 2.7.11, EC 2.7.11.1, Oncogene ARAF1, PKS, PKS2A-Raf proto-oncogene serine/threonine-protein kinase, Proto-oncogene A-Raf, Proto-oncogene A-Raf-1, Proto-oncogene Pks, RAFA1, Ras-binding protein DA-Raf, serine/threonine-protein kinase A-Raf, v-raf murine sarcoma 3611 viral oncogene homolog, v-raf murine sarcoma 3611 viral oncogene homolog 1
Gene Symbols ARAF
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 145-365 of human A-RAF (NP_001645) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 369.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.