Learn More
a-Ethyl-benzeneacetyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | alpha-Ethyl-benzeneacetyl Chloride |
| CAS | 36854-57-6 |
| Molecular Formula | C10H11ClO |
| Quantity | 5 g |
| Synonym | (±)-2-Phenylbutyric Chloride, (±)-alpha-Phenylbutyryl Chloride, 2-Phenylbutanoyl Chloride, 2-Phenylbutyryl Chloride, NSC 86133, alpha-Phenylbutyryl Chloride |
| InChI Formula | InChI=1S/C10H11ClO/c1-2-9(10(11)12)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3 |
| SMILES | CCC(C(=O)Cl)c1ccccc1 |
| IUPAC Name | 2-phenylbutanoyl chloride |
| Molecular Weight (g/mol) | 182.65 |
| Formula Weight | 182.0498 |