missing translation for 'onlineSavingsMsg'
Learn More

A-770041, MedChemExpress

Product Code. 16417077
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16417077

Brand: Medchem Express HY11011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-770041 is selective and orally active Src-family Lck inhibitor; A-770041 is a 147 nM inhibitor of Lck (1 mM ATP) and is 300-fold selective against Fyn, the other Src family kinase involved in T-cell signaling.

A-770041 is selective and orally active Src-family Lck inhibitor

Specifications

Chemical Name or Material A-770041
CAS 869748-10-7
Color White
Purity Grade Notes Research
Molecular Formula C34H39N9O3
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (160.84 mM)
SMILES NC1=C2C(N([C@@H]3CC[C@@H](N4CCN(C(C)=O)CC4)CC3)N=C2C5=CC=C(NC(C6=CC7=C(C=CC=C7)N6C)=O)C(OC)=C5)=NC=N1
Molecular Weight (g/mol) 621.73
Formula Weight 621.73
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.