missing translation for 'onlineSavingsMsg'
Learn More

A-381393, MedChemExpress

Product Code. 16443266
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443266

Brand: Medchem Express HY116941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-381393 is a potent, selective, brain penetrate dopamine D4 receptor antagonist, with Kis of 1.5, 1.9 and 1.6 nM for human dopamine D4.4, D4.2, and D4.7 receptor, respectively, >2700-fold selectivity over D1, D2, D3 and D5 dopamine receptors. A-381393 shows moderate affinity for 5-HT2A (Ki, 370 nM).

A-381393 is a potent, selective, brain penetrate dopamine D4 receptor antagonist, with Kis of 1.5, 1.9 and 1.6 nM for human dopamine D4.4, D4.2, and D4.7 receptor, respectively, >2700-fold selectivity over D1, D2, D3 and D5 dopamine receptors. A-381393 shows moderate affinity for 5-HT2A (Ki, 370 nM).

Specifications

Chemical Name or Material A-381393
Physical Form Solid
CAS 726174-00-1
Color Light Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C20H24N4
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 51.67 mg/mL (161.25 mM; Need ultrasonic)
SMILES CC1=CC=C(C=C1C)N2CCN(CC2)CC3=NC4=CC=CC=C4N3
Molecular Weight (g/mol) 320.43
Formula Weight 320.43
Percent Purity 99.9%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.