missing translation for 'onlineSavingsMsg'
Learn More

A-1165442, MedChemExpress

Product Code. 16485158
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485158

Brand: Medchem Express HY12428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-1165442 is a potent, competitive and orally available TRPV1 antagonist with an IC50 of 9 nM for human TRPV1.

A-1165442 is a potent, competitive and orally available TRPV1 antagonist with an IC50 of 9 nM for human TRPV1.

Specifications

Chemical Name or Material A-1165442
CAS 1221443-94-2
Color White
Purity Grade Notes Research
Molecular Formula C22H20ClF2N3O2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (231.56 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1=CC=CC2=C1C=C(C)N=C2)N[C@H]3C4=CC=C(Cl)C=C4OC(CF)(CF)C3
Molecular Weight (g/mol) 431.86
Formula Weight 431.86
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.