missing translation for 'onlineSavingsMsg'
Learn More

A-1155463, MedChemExpress

Product Code. 16417686
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16417686

Brand: Medchem Express HY19725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-1155463 is a highly potent and selective BCL-XL inhibitor with an EC50 of 70 nM in Molt-4 cell.

A-1155463 is a highly potent and selective BCL-XL inhibitor with an EC50 of 70 nM in Molt-4 cell.

Specifications

Chemical Name or Material A-1155463
CAS 1235034-55-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C35H32FN5O4S2
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (74.65 mM; Need ultrasonic)
SMILES O=C(C1=C(CCCOC2=CC=C(C#CCN(C)C)C=C2F)SC(N3CC4=C(C=CC=C4C(NC5=NC6=CC=CC=C6S5)=O)CC3)=N1)O
Molecular Weight (g/mol) 669.79
Formula Weight 669.79
Percent Purity 96.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.