Learn More
IKA™ A 10.4 Grinding Chamber Reduction
Product Code. 15829202 Shop All IKA Products
Click to view available options
:
For use with A 10 basic mill
This item is not returnable. View return policy
Product Code. 15829202
Brand: IKA™ 0025001664
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Designed for small grinding volumes and suitable for use in the A 10 basic mill.
  • For small grinding volumes
  • Stainless steel construction
  • Suitable for use with A 10 basic mill
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chamber Reduction
For Use With (Application) Grinding small volumes
For Use With (Equipment) A 10 basic mill
Material Stainless Steel